Learn More
Cayman Chemical Palmitelaidic Acid-d13, Purity: ≥99% deuterated forms (d1-d13), CAS: 2692623-91-7, 1 mg
SDP
Catalog No. 502940463
Encompass_Preferred
An internal standard for the quantification of palmitelaidic acid by GC- or LC-MS
Catalog No. 50-294-0463 Supplier Cayman Chemical Supplier No. 277131 MG
An internal standard for the quantification of palmitelaidic acid by GC- or LC-MS