Learn More
Medchemexpress LLC Palmitelaidic acid-d13 | 2692623-91-7 | 1 MG
SDP
Catalog No. 5000421049
Encompass_Preferred
Palmitelaidic acid-d13 is the deuterium-labeled form of Palmitelaidic Acid. Palmitelaidic Acid (9-trans-Hexadecenoic acid) is the trans isomer of palmitoleic acid. Palmitoleic acid is one of the most abundant fatty acids found in serum and tissue.
  • Can be used as a tracer
  • Used as an internal standard for quantitative analysis by methods such as NMR, GC-MS, or LC-MS
  • Stable heavy isotope of hydrogen incorporated into a drug molecule
  • Potential to influence pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-21049 Supplier Medchemexpress LLC Supplier No. HYN2341S1MG
Palmitelaidic acid-d13 is the deuterium-labeled form of Palmitelaidic Acid. Palmitelaidic Acid (9-trans-Hexadecenoic acid) is the trans isomer of palmitoleic acid. Palmitoleic acid is one of the most abundant fatty acids found in serum and tissue.
  • Can be used as a tracer
  • Used as an internal standard for quantitative analysis by methods such as NMR, GC-MS, or LC-MS
  • Stable heavy isotope of hydrogen incorporated into a drug molecule
  • Potential to influence pharmacokinetic and metabolic profiles of drugs