Learn More
Cayman Chemical Palmitic Acid-13C (C1 labeled), Purity: ≥98%, CAS: 57677-53-9, 250 mg
SDP
Catalog No. 502937279
Encompass_Preferred
An internal standard for the quantification of palmitic acid by GC- or LC-MS
Catalog No. 50-293-7279 Supplier Cayman Chemical Supplier No. 28749250 MG
An internal standard for the quantification of palmitic acid by GC- or LC-MS