Learn More
Cayman Chemical Palmitic Acid-13C (C2 labeled), Purity: ≥95%, CAS: 287100-87-2, 10 mg
SDP
Catalog No. 502939040
Encompass_Preferred
An internal standard for the quantification of palmitic acid by GC- or LC-MS
Catalog No. 50-293-9040 Supplier Cayman Chemical Supplier No. 2891610 MG
An internal standard for the quantification of palmitic acid by GC- or LC-MS