Learn More
Medchemexpress LLC Palmitic acid-1-13C | 57677-53-9 | 257.42 | 50 MG
SDP
Catalog No. 5000416280
Encompass_Preferred
Palmitic acid-1-13C is the 13C-labeled version of Palmitic acid. Palmitic acid is a long-chain saturated fatty acid naturally occurring in both animals and plants. It has been shown to induce the expression of glucose-regulated protein 78 (GRP78) and CCAAT/enhancer binding protein homologous protein (CHOP) in mouse granulosa cells.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-16280 Supplier Medchemexpress LLC Supplier No. HYN0830S350MG
Palmitic acid-1-13C is the 13C-labeled version of Palmitic acid. Palmitic acid is a long-chain saturated fatty acid naturally occurring in both animals and plants. It has been shown to induce the expression of glucose-regulated protein 78 (GRP78) and CCAAT/enhancer binding protein homologous protein (CHOP) in mouse granulosa cells.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.