Learn More
Cambridge Isotope Laboratories Palmitic Acid (1-13C, 99%), 57677-53-9, MFCD00002748, 1G
Catalog No. NC1462745
Encompass
Chemical Formula: CH3(CH2)14*COOH, Molecular Weight: 257.42, Synonym: Hexadecanoic acid-1-13C, MP: 61-64 °C.
Catalog No. NC1462745 Supplier Cambridge Isotope Laboratories Supplier No. CLM1501
Chemical Formula: CH3(CH2)14*COOH, Molecular Weight: 257.42, Synonym: Hexadecanoic acid-1-13C, MP: 61-64 °C.