Learn More
American Radiolabeled Chemicals Inc PALMITIC ACID-1-14C 50UC
SDP
Supplier:  American Radiolabeled Chemicals Inc ARC 0172A50UC
Encompass_Preferred
Palmitic acid-1-14C 50uCi in ethanol, 0.5 mCi/ml, S.A. 50-60 mCi/mmol, M.W. 256.4
Catalog No. 50-901-05529