Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Palmitic Acid, [1-14C], MP Biomedicals™

Catalog No. mp011219550 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 mci; 37 mbq
250 μci; 9.25 mbq
50 μci; 1.85 mbq
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP011219550 50 μci; 1.85 mbq
MP011219583 250 μci; 9.25 mbq
MP011219591 1 mci; 37 mbq
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MP011219550 Supplier MP Biomedicals, Inc Supplier No. 011219550

Molecular Formula CH3(CH2)14COOH

  • Sp. Act. 40-60 mCi/mmol; 1.48-2.22 GBq/mmol
  • Ethanol
TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula CH3(CH2)14COOH

Specifications

Quantity 50 μci; 1.85 mbq
storageNote1 Store at 2-8°C.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.