Learn More
Medchemexpress LLC Palmitic acid-1,2,3,4-13C4 | 287100-89-4 | 260.39 | 25 MG
SDP
Catalog No. 5000389386
Encompass_Preferred
Palmitic acid-1,2,3,4-13C4 is the 13C-labeled Palmitic acid, a long-chain saturated fatty acid found in animals and plants. It can induce the expression of glucose-regulated protein 78 (GRP78) and CCAAT/enhancer binding protein homologous protein (CHOP) in mouse granulosa cells. It is for research use only.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-003-89386 Supplier Medchemexpress LLC Supplier No. 25MGHYN0830S
Palmitic acid-1,2,3,4-13C4 is the 13C-labeled Palmitic acid, a long-chain saturated fatty acid found in animals and plants. It can induce the expression of glucose-regulated protein 78 (GRP78) and CCAAT/enhancer binding protein homologous protein (CHOP) in mouse granulosa cells. It is for research use only.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS