Learn More
Medchemexpress LLC Palmitic acid-1,2,3,4-13C4 | 287100-89-4 | 260.39 | 50 MG
SDP
Catalog No. 5000417014
Encompass_Preferred
Palmitic acid-1,2,3,4-13C4 is a 13C-labeled version of palmitic acid, a long-chain saturated fatty acid found in both animals and plants. It has been shown to induce the expression of glucose-regulated protein 78 (GRP78) and CCAAT/enhancer binding protein homologous protein (CHOP) in mouse granulosa cells.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-17014 Supplier Medchemexpress LLC Supplier No. HYN0830S50MG
Palmitic acid-1,2,3,4-13C4 is a 13C-labeled version of palmitic acid, a long-chain saturated fatty acid found in both animals and plants. It has been shown to induce the expression of glucose-regulated protein 78 (GRP78) and CCAAT/enhancer binding protein homologous protein (CHOP) in mouse granulosa cells.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.