Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Palmitic acid, 1,2,3,4-13C4 | 287100-89-4 | 99.0% | 260.39 g/mol | C12(13C)4H32O2 | 10 MG
SDP

Catalog No. 5000296203
Encompass_Preferred

Palmitic acid labeled with four 13C atoms at positions 1-4, used as a stable isotope tracer for metabolic and analytical studies. It is a long-chain saturated fatty acid prepared for isotopic labeling applications.

  • 13C-labeled at positions 1-4.
  • Purity 99.0%.
  • CAS number 287100-89-4.
  • Molecular weight 260.39 g/mol.
  • Supplied as a 10 mg vial for tracer and analytical use.

Catalog No. 50-002-96203 Supplier Medchemexpress LLC Supplier No. HYN0830S10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Palmitic acid labeled with four 13C atoms at positions 1-4, used as a stable isotope tracer for metabolic and analytical studies. It is a long-chain saturated fatty acid prepared for isotopic labeling applications.

  • 13C-labeled at positions 1-4.
  • Purity 99.0%.
  • CAS number 287100-89-4.
  • Molecular weight 260.39 g/mol.
  • Supplied as a 10 mg vial for tracer and analytical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.