Learn More
Medchemexpress LLC Palmitic acid-13C | 287100-87-2 | 257.42 | 25 MG
SDP
Catalog No. 5000390152
Encompass_Preferred
Palmitic acid-13C is the 13C-labeled Palmitic acid. It is a long-chain saturated fatty acid commonly found in both animals and plants. It can induce the expression of glucose-regulated protein 78 (GRP78) and CCAAT/enhancer binding protein homologous protein (CHOP) in mouse granulosa cells.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-003-90152 Supplier Medchemexpress LLC Supplier No. 25MGHYN0830S9
Palmitic acid-13C is the 13C-labeled Palmitic acid. It is a long-chain saturated fatty acid commonly found in both animals and plants. It can induce the expression of glucose-regulated protein 78 (GRP78) and CCAAT/enhancer binding protein homologous protein (CHOP) in mouse granulosa cells.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS