Learn More
Medchemexpress LLC Palmitic acid-13C | 287100-87-2 | 257.42 | 50 MG
SDP
Catalog No. 5000416930
Encompass_Preferred
Palmitic acid-13C is the 13C-labeled version of Palmitic acid. This long-chain saturated fatty acid is found in both animals and plants and can induce the expression of glucose-regulated protein 78 (GRP78) and CCAAT/enhancer binding protein homologous protein (CHOP) in mouse granulosa cells.
  • Can be used as a tracer.
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-16930 Supplier Medchemexpress LLC Supplier No. HYN0830S950MG
Palmitic acid-13C is the 13C-labeled version of Palmitic acid. This long-chain saturated fatty acid is found in both animals and plants and can induce the expression of glucose-regulated protein 78 (GRP78) and CCAAT/enhancer binding protein homologous protein (CHOP) in mouse granulosa cells.
  • Can be used as a tracer.
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.