Learn More
Medchemexpress LLC Palmitic acid-13C16 | 56599-85-0 | 272.31 | 100 MG
SDP
Catalog No. 5000417079
Encompass_Preferred
Palmitic acid-13C16 is a 13C-labeled version of Palmitic acid, a long-chain saturated fatty acid found in animals and plants. It has been shown to induce expression of glucose-regulated protein 78 (GRP78) and CCAAT/enhancer binding protein homologous protein (CHOP) in mouse granulosa cells. This product is for research use only and not for patient use.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotopes incorporated for quantitation during drug development
Catalog No. 50-004-17079 Supplier Medchemexpress LLC Supplier No. HYN0830S6100MG
Palmitic acid-13C16 is a 13C-labeled version of Palmitic acid, a long-chain saturated fatty acid found in animals and plants. It has been shown to induce expression of glucose-regulated protein 78 (GRP78) and CCAAT/enhancer binding protein homologous protein (CHOP) in mouse granulosa cells. This product is for research use only and not for patient use.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotopes incorporated for quantitation during drug development