Learn More
Medchemexpress LLC Palmitic acid-13C16 | 56599-85-0 | 272.31 | 25 MG
SDP
Catalog No. 5000389384
Encompass_Preferred
Palmitic acid-13C16 is the 13C-labeled Palmitic acid, a long-chain saturated fatty acid found in animals and plants. It can induce the expression of glucose-regulated protein 78 (GRP78) and CCAAT/enhancer binding protein homologous protein (CHOP) in mouse granulosa cells. This compound can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Stable heavy isotopes are incorporated into drug molecules as tracers for quantitation during drug development, with deuteration potentially affecting pharmacokinetic and metabolic profiles.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Incorporates stable heavy isotopes into drug molecules for quantitation
  • Deuteration may affect pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-003-89384 Supplier Medchemexpress LLC Supplier No. 25MGHYN0830S6
Palmitic acid-13C16 is the 13C-labeled Palmitic acid, a long-chain saturated fatty acid found in animals and plants. It can induce the expression of glucose-regulated protein 78 (GRP78) and CCAAT/enhancer binding protein homologous protein (CHOP) in mouse granulosa cells. This compound can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Stable heavy isotopes are incorporated into drug molecules as tracers for quantitation during drug development, with deuteration potentially affecting pharmacokinetic and metabolic profiles.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Incorporates stable heavy isotopes into drug molecules for quantitation
  • Deuteration may affect pharmacokinetic and metabolic profiles of drugs