Learn More
Medchemexpress LLC Palmitic acid-13C16 | 56599-85-0 | 272.31 | 5 MG
SDP
Catalog No. 5000416962
Encompass_Preferred
Palmitic acid-13C16 is a 13C-labeled version of Palmitic acid, a long-chain saturated fatty acid found in animals and plants. It can induce the expression of glucose-regulated protein 78 (GRP78) and CCAAT/enhancer binding protein homologous protein (CHOP) in mouse granulosa cells.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-16962 Supplier Medchemexpress LLC Supplier No. HYN0830S65MG
Palmitic acid-13C16 is a 13C-labeled version of Palmitic acid, a long-chain saturated fatty acid found in animals and plants. It can induce the expression of glucose-regulated protein 78 (GRP78) and CCAAT/enhancer binding protein homologous protein (CHOP) in mouse granulosa cells.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS