Learn More
Medchemexpress LLC Palmitic acid-13C16 | 56599-85-0 | 99.4% | 10 MG
SDP
Catalog No. 5000296202
Encompass_Preferred
Palmitic acid-13C16 is the uniformly 13C-labeled form of palmitic (hexadecanoic) acid used as a stable isotope tracer and internal standard for mass spectrometry, metabolic tracing, and lipidomics. It preserves the chemical properties of native palmitic acid while providing a distinct mass shift for analytical detection.
  • Uniformly 13C-labeled (16 carbons) for clear mass spectrometry separation.
  • Suitable as an internal standard for quantitative lipidomics and metabolic tracing.
  • High isotopic and chemical purity for reliable analytical results.
  • Supplied as a small solid sample convenient for laboratory use.
  • Compatible with common extraction and derivatization procedures.
Catalog No. 50-002-96202 Supplier Medchemexpress LLC Supplier No. HYN0830S610MG
Palmitic acid-13C16 is the uniformly 13C-labeled form of palmitic (hexadecanoic) acid used as a stable isotope tracer and internal standard for mass spectrometry, metabolic tracing, and lipidomics. It preserves the chemical properties of native palmitic acid while providing a distinct mass shift for analytical detection.
  • Uniformly 13C-labeled (16 carbons) for clear mass spectrometry separation.
  • Suitable as an internal standard for quantitative lipidomics and metabolic tracing.
  • High isotopic and chemical purity for reliable analytical results.
  • Supplied as a small solid sample convenient for laboratory use.
  • Compatible with common extraction and derivatization procedures.