Learn More
Medchemexpress LLC Palmitic acid-13C16 sodium | 2483736-17-8 | 294.29 | 1 MG
SDP
Catalog No. 5000417093
Encompass_Preferred
Palmitic acid-13C16 sodium is a 13C-labeled, long-chain saturated fatty acid. Commonly found in both animals and plants, this compound can induce the expression of glucose-regulated protein 78 (GRP78) and CCAAT/enhancer binding protein homologous protein (CHOP) in mouse granulosa cells.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-17093 Supplier Medchemexpress LLC Supplier No. HYN0830BS11MG
Add to Cart
Add to Cart
Palmitic acid-13C16 sodium is a 13C-labeled, long-chain saturated fatty acid. Commonly found in both animals and plants, this compound can induce the expression of glucose-regulated protein 78 (GRP78) and CCAAT/enhancer binding protein homologous protein (CHOP) in mouse granulosa cells.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.