Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Palmitic acid-13C16 sodium | 2483736-17-8 | MFCD11898946 | 99.1% | 294.29 g/mol | 13C16H31NaO2 | 10 MG
SDP

Catalog No. 5000295990
Encompass_Preferred

Palmitic acid-13C16 sodium is the sodium salt of palmitic acid in which all 16 carbon atoms are isotopically labeled with carbon-13. It is supplied as a research-grade stable isotope tracer and internal standard for quantitative analysis by NMR, GC-MS, and LC-MS. For research use only; not for human or clinical use.

  • Isotopically labeled: all 16 carbons are 13C-labeled, enabling clear mass shifts for tracer studies.
  • Sodium salt form: improved handling and solubility compared with the free acid for certain workflows.
  • High purity: analytical grade purity suitable for quantitative standards and method development.
  • Small pack sizes available: sold in milligram quantities for low-consumption analytical applications.
  • Analytical compatibility: compatible with NMR, GC-MS, and LC-MS quantitation workflows.

Catalog No. 50-002-95990 Supplier Medchemexpress LLC Supplier No. HYN0830BS110MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Palmitic acid-13C16 sodium is the sodium salt of palmitic acid in which all 16 carbon atoms are isotopically labeled with carbon-13. It is supplied as a research-grade stable isotope tracer and internal standard for quantitative analysis by NMR, GC-MS, and LC-MS. For research use only; not for human or clinical use.

  • Isotopically labeled: all 16 carbons are 13C-labeled, enabling clear mass shifts for tracer studies.
  • Sodium salt form: improved handling and solubility compared with the free acid for certain workflows.
  • High purity: analytical grade purity suitable for quantitative standards and method development.
  • Small pack sizes available: sold in milligram quantities for low-consumption analytical applications.
  • Analytical compatibility: compatible with NMR, GC-MS, and LC-MS quantitation workflows.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.