Learn More
Medchemexpress LLC Palmitic acid-13C2 | 86683-25-2 | 258.41 | 100 MG
SDP
Catalog No. 5000416926
Encompass_Preferred
Palmitic acid-13C2 is a 13C-labeled long-chain saturated fatty acid commonly found in animals and plants. It can induce the expression of glucose-regulated protein 78 (GRP78) and CCAAT/enhancer binding protein homologous protein (CHOP) in mouse granulosa cells. This product can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Tracer for metabolic studies
  • Internal standard for quantitative analysis
  • Induces expression of GRP78 and CHOP in mouse granulosa cells
Catalog No. 50-004-16926 Supplier Medchemexpress LLC Supplier No. HYN0830S10100MG
Palmitic acid-13C2 is a 13C-labeled long-chain saturated fatty acid commonly found in animals and plants. It can induce the expression of glucose-regulated protein 78 (GRP78) and CCAAT/enhancer binding protein homologous protein (CHOP) in mouse granulosa cells. This product can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Tracer for metabolic studies
  • Internal standard for quantitative analysis
  • Induces expression of GRP78 and CHOP in mouse granulosa cells