Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Palmitic acid-13C2 | 86683-25-2 | 258.41 | 25 MG
SDP

Catalog No. 5000390307
Encompass_Preferred

Palmitic acid-13C2 is a 13C-labeled version of Palmitic acid, a long-chain saturated fatty acid found in animals and plants. It has been observed to induce the expression of glucose-regulated protein 78 (GRP78) and CCAAT/enhancer binding protein homologous protein (CHOP) in mouse granulosa cells.

  • Purity: 99.54%
  • Intended for research use only
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Powder is stable for 3 years at -20°C and 2 years at 4°C
  • In solvent, stable for 6 months at -80°C and 1 month at -20°C
  • Soluble in DMSO (≥ 100 mg/mL)

Catalog No. 50-003-90307 Supplier Medchemexpress LLC Supplier No. 25MGHYN0830S10
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Palmitic acid-13C2 is a 13C-labeled version of Palmitic acid, a long-chain saturated fatty acid found in animals and plants. It has been observed to induce the expression of glucose-regulated protein 78 (GRP78) and CCAAT/enhancer binding protein homologous protein (CHOP) in mouse granulosa cells.

  • Purity: 99.54%
  • Intended for research use only
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Powder is stable for 3 years at -20°C and 2 years at 4°C
  • In solvent, stable for 6 months at -80°C and 1 month at -20°C
  • Soluble in DMSO (≥ 100 mg/mL)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.