Learn More
Cayman Chemical PalmItIc AcId-13CC1 labeled 1g
SDP
Catalog No. 502937278
Encompass_Preferred
An internal standard for the quantification of palmitic acid by GC- or LC-MS
Catalog No. 50-293-7278 Supplier Cayman Chemical Supplier No. 287491 G
An internal standard for the quantification of palmitic acid by GC- or LC-MS