Learn More
Cayman Chemical Palmitic Acid-13C (C1, C2, C3, and C4 labeled), Purity: ≥98%, CAS: 287100-89-4, 100 mg
SDP
Catalog No. 502991809
Encompass_Preferred
An internal standard for the quantification of palmitic acid by GC- or LC-MS
Catalog No. 50-299-1809 Supplier Cayman Chemical Supplier No. 30550100 MG
An internal standard for the quantification of palmitic acid by GC- or LC-MS