Learn More
Cayman Chemical Palmitic Acid-13C (C1, C2, C3, and C4 labeled), Purity: ≥98%, CAS: 287100-89-4, 25 mg
SDP
Catalog No. 502992189
Encompass_Preferred
An internal standard for the quantification of palmitic acid by GC- or LC-MS
Catalog No. 50-299-2189 Supplier Cayman Chemical Supplier No. 3055025 MG
An internal standard for the quantification of palmitic acid by GC- or LC-MS