Learn More
Cayman Chemical PalmItIc AcId-13CC1C2C3 50mg
SDP
Catalog No. 502991313
Encompass_Preferred
An internal standard for the quantification of palmitic acid by GC- or LC-MS
Catalog No. 50-299-1313 Supplier Cayman Chemical Supplier No. 3055050 MG
An internal standard for the quantification of palmitic acid by GC- or LC-MS