Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Palmitic acid-2-13C | 287100-87-2 | MFCD00083983 | 98.0% | 10 MG
SDP

Catalog No. 5000296562
Encompass_Preferred

Palmitic acid-13C is a stable isotope-labeled palmitic acid supplied for use as a tracer or internal standard in analytical and biochemical applications, including GC-MS and LC-MS workflows. It provides a defined isotopic label for metabolic tracing and quantification.

  • Used as a tracer and internal standard in GC-MS and LC-MS analyses.
  • Stable isotope labeled (13C) fatty acid for metabolic tracing.
  • Purity 98.0% as provided.
  • CAS number 287100-87-2 for unambiguous identification.
  • Provided as a 10 MG package; additional sizes available.

Catalog No. 50-002-96562 Supplier Medchemexpress LLC Supplier No. HYN0830S910MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Palmitic acid-13C is a stable isotope-labeled palmitic acid supplied for use as a tracer or internal standard in analytical and biochemical applications, including GC-MS and LC-MS workflows. It provides a defined isotopic label for metabolic tracing and quantification.

  • Used as a tracer and internal standard in GC-MS and LC-MS analyses.
  • Stable isotope labeled (13C) fatty acid for metabolic tracing.
  • Purity 98.0% as provided.
  • CAS number 287100-87-2 for unambiguous identification.
  • Provided as a 10 MG package; additional sizes available.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.