Learn More
U.S. Pharmacopeia Palmitic Acid, 57-10-3, MFCD00002747, 500mg
Catalog No. NC9094442
Encompass
Molecular formula C16H32O2, Molecular weight 256.42, Melting Point 132.8 - 147.2 °F (56 - 64 °C), Boiling Point 664.7 °F (351.5 °C), Synonyms: Cetylic acid, 1-Pentadecanecarboxylic acid
Catalog No. NC9094442 Supplier U.S. Pharmacopeia Supplier No. 1492007
Molecular formula C16H32O2, Molecular weight 256.42, Melting Point 132.8 - 147.2 °F (56 - 64 °C), Boiling Point 664.7 °F (351.5 °C), Synonyms: Cetylic acid, 1-Pentadecanecarboxylic acid