Learn More
Medchemexpress LLC Palmitic acid | 57-10-3 | 256.42 | 10 G
SDP
Catalog No. 5000416762
Encompass_Preferred
Palmitic acid is a long-chain saturated fatty acid found in both animals and plants. It can induce the expression of glucose-regulated protein 78 (GRP78) and CCAAT/enhancer binding protein homologous protein (CHOP) in mouse granulosa cells. Palmitic acid is also used to establish a cell steatosis model.
  • Long-chain saturated fatty acid
  • Found in both animals and plants
  • Induces expression of GRP78 and CHOP in mouse granulosa cells
  • Used to establish a cell steatosis model
  • Purity: 99.86%
Catalog No. 50-004-16762 Supplier Medchemexpress LLC Supplier No. HYN083010G
Palmitic acid is a long-chain saturated fatty acid found in both animals and plants. It can induce the expression of glucose-regulated protein 78 (GRP78) and CCAAT/enhancer binding protein homologous protein (CHOP) in mouse granulosa cells. Palmitic acid is also used to establish a cell steatosis model.
  • Long-chain saturated fatty acid
  • Found in both animals and plants
  • Induces expression of GRP78 and CHOP in mouse granulosa cells
  • Used to establish a cell steatosis model
  • Purity: 99.86%