Learn More
Cayman Chemical PalmItIc AcId Alkyn 10mg
SDP
Catalog No. 502952787
Encompass_Preferred
A form of palmitic acid with an ω-terminal alkyne for use in click chemistry to isolate palmitoylated proteins
Catalog No. 50-295-2787 Supplier Cayman Chemical Supplier No. 1326610 MG
A form of palmitic acid with an ω-terminal alkyne for use in click chemistry to isolate palmitoylated proteins