Learn More
Cayman Chemical Palmitic Acid-d2, Purity: ≥98% deuterated forms (d1-d2), CAS: 62689-96-7, 250 mg
SDP
Catalog No. 502893848
Encompass_Preferred
An internal standard for the quantification of palmitic acid by GC- or LC-MS
Catalog No. 50-289-3848 Supplier Cayman Chemical Supplier No. 9001876250 MG
An internal standard for the quantification of palmitic acid by GC- or LC-MS