Learn More
Medchemexpress LLC Palmitic acid-d2 | 62689-96-7 | 258.44 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYN0830S4
Encompass_Preferred
Palmitic acid-d2 is a deuterium-labeled palmitic acid, which is a long-chain saturated fatty acid found in both animals and plants. It can induce the expression of glucose-regulated protein 78 (GRP78) and CCAAT/enhancer binding protein homologous protein (CHOP) in mouse granulosa cells.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-003-89249