Learn More
Medchemexpress LLC Palmitic acid-d2 | 62689-96-7 | 258.44 | 50 MG
SDP
Catalog No. 5000416960
Encompass_Preferred
Palmitic acid-d2 is the deuterium labeled Palmitic acid. Palmitic acid is a long-chain saturated fatty acid commonly found in both animals and plants. Palmitic acid can induce the expression of glucose-regulated protein 78 (GRP78) and CCAAT/enhancer binding protein homologous protein (CHOP) in mouse granulosa cells. It can be used as a tracer or as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuterium labeled palmitic acid.
  • Long-chain saturated fatty acid.
  • Found in both animals and plants.
  • Induces expression of glucose-regulated protein 78 (GRP78) and CCAAT/enhancer binding protein homologous protein (CHOP) in mouse granulosa cells.
  • Can be used as a tracer or an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-16960 Supplier Medchemexpress LLC Supplier No. HYN0830S450MG
Palmitic acid-d2 is the deuterium labeled Palmitic acid. Palmitic acid is a long-chain saturated fatty acid commonly found in both animals and plants. Palmitic acid can induce the expression of glucose-regulated protein 78 (GRP78) and CCAAT/enhancer binding protein homologous protein (CHOP) in mouse granulosa cells. It can be used as a tracer or as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuterium labeled palmitic acid.
  • Long-chain saturated fatty acid.
  • Found in both animals and plants.
  • Induces expression of glucose-regulated protein 78 (GRP78) and CCAAT/enhancer binding protein homologous protein (CHOP) in mouse granulosa cells.
  • Can be used as a tracer or an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.