Learn More
Medchemexpress LLC Palmitic acid-d31 | 39756-30-4 | 287.62 | 100 MG
SDP
Catalog No. 5000416361
Encompass_Preferred
Palmitic acid-d31 is the deuterium labeled Palmitic acid, which is a long-chain saturated fatty acid commonly found in both animals and plants. It can induce the expression of glucose-regulated protein 78 (GRP78) and CCAAT/enhancer binding protein homologous protein (CHOP) in mouse granulosa cells.
  • High purity (99.56%).
  • Solid, white to off-white appearance.
  • Used as a tracer.
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuteration can affect the pharmacokinetic and metabolic profiles of drugs.
  • Induces the expression of glucose-regulated protein 78 (GRP78) and CCAAT/enhancer binding protein homologous protein (CHOP) in mouse granulosa cells.
Catalog No. 50-004-16361 Supplier Medchemexpress LLC Supplier No. HYN0830S2100MG
Palmitic acid-d31 is the deuterium labeled Palmitic acid, which is a long-chain saturated fatty acid commonly found in both animals and plants. It can induce the expression of glucose-regulated protein 78 (GRP78) and CCAAT/enhancer binding protein homologous protein (CHOP) in mouse granulosa cells.
  • High purity (99.56%).
  • Solid, white to off-white appearance.
  • Used as a tracer.
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuteration can affect the pharmacokinetic and metabolic profiles of drugs.
  • Induces the expression of glucose-regulated protein 78 (GRP78) and CCAAT/enhancer binding protein homologous protein (CHOP) in mouse granulosa cells.