Learn More
Medchemexpress LLC Palmitic acid-d31 | 39756-30-4 | 287.62 | 5 MG
SDP
Catalog No. 5000417065
Encompass_Preferred
Palmitic acid-d31 is a deuterium-labeled palmitic acid, a long-chain saturated fatty acid commonly found in both animals and plants. It can induce glucose-regulated protein 78 (GRP78) and CCAAT/enhancer binding protein homologous protein (CHOP) in mouse granulosa cells. This compound is valuable as a tracer or an internal standard for quantitative analysis in various spectroscopic methods, and its deuterium labeling offers potential benefits for studying pharmacokinetic and metabolic profiles.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuterium labeling for pharmacokinetic and metabolic studies
Catalog No. 50-004-17065 Supplier Medchemexpress LLC Supplier No. HYN0830BS35MG
Palmitic acid-d31 is a deuterium-labeled palmitic acid, a long-chain saturated fatty acid commonly found in both animals and plants. It can induce glucose-regulated protein 78 (GRP78) and CCAAT/enhancer binding protein homologous protein (CHOP) in mouse granulosa cells. This compound is valuable as a tracer or an internal standard for quantitative analysis in various spectroscopic methods, and its deuterium labeling offers potential benefits for studying pharmacokinetic and metabolic profiles.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuterium labeling for pharmacokinetic and metabolic studies