Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Palmitic acid-d31 | 39756-30-4 | 287.62 | 500 MG
SDP

Catalog No. 5000417016
Encompass_Preferred

Palmitic acid-d31 is the deuterium labeled Palmitic acid. Palmitic acid is a long-chain saturated fatty acid commonly found in both animals and plants. It can induce the expression of glucose-regulated protein 78 (GRP78) and CCAAT/enhancer binding protein homologous protein (CHOP) in mouse granulosa cells.

  • Deuterium labeled palmitic acid.
  • Long-chain saturated fatty acid.
  • Commonly found in both animals and plants.
  • Can induce the expression of glucose-regulated protein 78 (GRP78) and CCAAT/enhancer binding protein homologous protein (CHOP) in mouse granulosa cells.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Purity: 99.56%.
  • Appearance: Solid.
  • Color: White to off-white.

Catalog No. 50-004-17016 Supplier Medchemexpress LLC Supplier No. HYN0830S2500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Palmitic acid-d31 is the deuterium labeled Palmitic acid. Palmitic acid is a long-chain saturated fatty acid commonly found in both animals and plants. It can induce the expression of glucose-regulated protein 78 (GRP78) and CCAAT/enhancer binding protein homologous protein (CHOP) in mouse granulosa cells.

  • Deuterium labeled palmitic acid.
  • Long-chain saturated fatty acid.
  • Commonly found in both animals and plants.
  • Can induce the expression of glucose-regulated protein 78 (GRP78) and CCAAT/enhancer binding protein homologous protein (CHOP) in mouse granulosa cells.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Purity: 99.56%.
  • Appearance: Solid.
  • Color: White to off-white.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.