Learn More
Medchemexpress LLC Palmitic acid-d4-1 | 75736-47-9 | 260.45 | 1 MG
SDP
Catalog No. 5000416958
Encompass_Preferred
Palmitic acid-d4-1 is a chemical compound commonly used in laboratory research and for the manufacture of various substances. It is supplied as a solid with a melting point range of 60-63°C. This product requires careful handling due to potential health hazards including skin and eye irritation, and it may cause respiratory irritation if inhaled. It is also harmful if swallowed.
  • Harmful if swallowed
  • Causes skin irritation
  • Causes serious eye irritation
  • May cause respiratory irritation
  • Solid appearance
  • Melting/freezing point 60-63°C
  • Store powder at -20°C for 3 years or 4°C for 2 years
  • Store in solvent at -80°C for 6 months or -20°C for 1 month
Catalog No. 50-004-16958 Supplier Medchemexpress LLC Supplier No. HYN0830S121MG
Palmitic acid-d4-1 is a chemical compound commonly used in laboratory research and for the manufacture of various substances. It is supplied as a solid with a melting point range of 60-63°C. This product requires careful handling due to potential health hazards including skin and eye irritation, and it may cause respiratory irritation if inhaled. It is also harmful if swallowed.
  • Harmful if swallowed
  • Causes skin irritation
  • Causes serious eye irritation
  • May cause respiratory irritation
  • Solid appearance
  • Melting/freezing point 60-63°C
  • Store powder at -20°C for 3 years or 4°C for 2 years
  • Store in solvent at -80°C for 6 months or -20°C for 1 month