Learn More
Medchemexpress LLC Palmitic acid-d4 | 75736-49-1 | 260.45 | 5 MG
SDP
Catalog No. 5000417080
Encompass_Preferred
Palmitic acid-d4 is a deuterium-labeled version of palmitic acid, a long-chain saturated fatty acid commonly found in both animals and plants. It has been shown to induce the expression of glucose-regulated protein 78 (GRP78) and CCAAT/enhancer binding protein homologous protein (CHOP) in mouse granulosa cells.
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-17080 Supplier Medchemexpress LLC Supplier No. HYN0830S75MG
Palmitic acid-d4 is a deuterium-labeled version of palmitic acid, a long-chain saturated fatty acid commonly found in both animals and plants. It has been shown to induce the expression of glucose-regulated protein 78 (GRP78) and CCAAT/enhancer binding protein homologous protein (CHOP) in mouse granulosa cells.
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS