Learn More
Medchemexpress LLC Palmitic acid-d4 | 75736-49-1 | 260.45 | 50 MG
SDP
Catalog No. 5000416261
Encompass_Preferred
Palmitic acid-d4 is the deuterium labeled Palmitic acid, a long-chain saturated fatty acid found in animals and plants. It can induce the expression of glucose-regulated protein 78 (GRP78) and CCAAT/enhancer binding protein homologous protein (CHOP) in mouse granulosa cells.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-16261 Supplier Medchemexpress LLC Supplier No. HYN0830S750MG
Palmitic acid-d4 is the deuterium labeled Palmitic acid, a long-chain saturated fatty acid found in animals and plants. It can induce the expression of glucose-regulated protein 78 (GRP78) and CCAAT/enhancer binding protein homologous protein (CHOP) in mouse granulosa cells.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.