Learn More
Cayman Chemical PalmItIc AcId-d9 500ug
SDP
Catalog No. 502915519
Encompass_Preferred
An internal standard for the quantification of palmitic acid by GC- or LC-MS
Catalog No. 50-291-5519 Supplier Cayman Chemical Supplier No. 37061500 UG
An internal standard for the quantification of palmitic acid by GC- or LC-MS