Learn More
Cayman Chemical Palmitic Acid ethyl ester, Purity: ≥98%, CAS: 628-97-7, 25 g
SDP
Catalog No. 502891178
Encompass_Preferred
Palmitic acid ethyl ester is a neutral, lipid-soluble form of the free acid. It is one of the fatty acid ethyl esters that increase cytosolic Ca2+ concentration leading to pancreatic acinar cell injury due to excessive consumption of ethanol.{14120}
Catalog No. 50-289-1178 Supplier Cayman Chemical Supplier No. 1000820225 G
Palmitic acid ethyl ester is a neutral, lipid-soluble form of the free acid. It is one of the fatty acid ethyl esters that increase cytosolic Ca2+ concentration leading to pancreatic acinar cell injury due to excessive consumption of ethanol.{14120}