Learn More
Cayman Chemical Palmitic Acid ethyl ester
SDP
Supplier:  Cayman Chemical 1000820210 G
Encompass_Preferred
Palmitic acid ethyl ester is a neutral, lipid-soluble form of the free acid. It is one of the fatty acid ethyl esters that increase cytosolic Ca2+ concentration leading to pancreatic acinar cell injury due to excessive consumption of ethanol.{14120}
Catalog No. 50-288-9563