Learn More
Cayman Chemical Palmitic Acid methyl ester
SDP
Supplier:  Cayman Chemical 1000735825 G
Encompass_Preferred
A saturated fatty acid methyl ester; concentration in cells is modulated by methanol; inhibits phagocytosis and decreases cell viability in isolated Kupffer cells; decreases secretion of IL-10, TNF-α, NO, and PGE2 in LPS-treated cells
Catalog No. 50-288-8748