Learn More
Cayman Chemical Palmitic Acid methyl ester, Purity: ≥99%, CAS: 112-39-0, 10 g
SDP
Catalog No. 502889454
Encompass_Preferred
A saturated fatty acid methyl ester; concentration in cells is modulated by methanol; inhibits phagocytosis and decreases cell viability in isolated Kupffer cells; decreases secretion of IL-10, TNF-α, NO, and PGE2 in LPS-treated cells
Catalog No. 50-288-9454 Supplier Cayman Chemical Supplier No. 1000735810 G
A saturated fatty acid methyl ester; concentration in cells is modulated by methanol; inhibits phagocytosis and decreases cell viability in isolated Kupffer cells; decreases secretion of IL-10, TNF-α, NO, and PGE2 in LPS-treated cells