Learn More
Cayman Chemical Palmitic Acid methyl ester, Purity: ≥99%, CAS: 112-39-0, 5 g
SDP
Catalog No. 502890948
Encompass_Preferred
A saturated fatty acid methyl ester; concentration in cells is modulated by methanol; inhibits phagocytosis and decreases cell viability in isolated Kupffer cells; decreases secretion of IL-10, TNF-α, NO, and PGE2 in LPS-treated cells
Catalog No. 50-289-0948 Supplier Cayman Chemical Supplier No. 100073585 G
A saturated fatty acid methyl ester; concentration in cells is modulated by methanol; inhibits phagocytosis and decreases cell viability in isolated Kupffer cells; decreases secretion of IL-10, TNF-α, NO, and PGE2 in LPS-treated cells