Learn More
Medchemexpress LLC Palmitic acid N-hydroxysuccinimide | 14464-31-4 | 98.0% | 353.50 | 1 G
SDP
Catalog No. 5000451478
Encompass_Preferred
Palmitic acid N-hydroxysuccinimide is used to conjugate proteins to prepare targeted delivery vectors. It can also be used for the covalent connection between palmitic acid and ovalbumin, and to synthesize various amino and thiogenic compounds.
  • Functions as a lipophilic electrophile.
  • Suitable for covalent connection between palmitic acid and ovalbumin.
  • Used to synthesize cetacyl derivatives of amino acids.
  • Used to synthesize aminoacyl-tRNA.
  • Used to synthesize coenzyme A.
  • Used to synthesize mercaptoacetic acid.
  • Used to synthesize other amino and thiogenic compounds.
Catalog No. 50-004-51478 Supplier Medchemexpress LLC Supplier No. HYW0111341G
Palmitic acid N-hydroxysuccinimide is used to conjugate proteins to prepare targeted delivery vectors. It can also be used for the covalent connection between palmitic acid and ovalbumin, and to synthesize various amino and thiogenic compounds.
  • Functions as a lipophilic electrophile.
  • Suitable for covalent connection between palmitic acid and ovalbumin.
  • Used to synthesize cetacyl derivatives of amino acids.
  • Used to synthesize aminoacyl-tRNA.
  • Used to synthesize coenzyme A.
  • Used to synthesize mercaptoacetic acid.
  • Used to synthesize other amino and thiogenic compounds.