Learn More
Nanocs Palmitic acid PEG, mPEG-PLA, Specification: 5000 Da,100 mg
Catalog No. NC2810947
Encompass
Palmitic acid is a 16 carbon saturated fatty acid lipid with excellent hydrophobicity. PEG modified palmitic acid is an excellent amphilphilic polymer with both hydrophilicity and hydrophobility. Pegylated lipids are excellent liposome formation materials that can be used for drug delivery, gene transfection and vaccine delivery as well. These materials can also be used for targeted drug delivery by modifying their surfaces with targeting ligands such as antibodies, peptide
Catalog No. NC2810947 Supplier Nanocs Supplier No. PG1PLA5K
Palmitic acid is a 16 carbon saturated fatty acid lipid with excellent hydrophobicity. PEG modified palmitic acid is an excellent amphilphilic polymer with both hydrophilicity and hydrophobility. Pegylated lipids are excellent liposome formation materials that can be used for drug delivery, gene transfection and vaccine delivery as well. These materials can also be used for targeted drug delivery by modifying their surfaces with targeting ligands such as antibodies, peptide