Learn More
Broadpharm Palmitic acid-PEG4-NHS ester, 100mg
Catalog No. NC4036241
Encompass
Palmitic acid-PEG4-NHS ester is a saturated long-chain fatty acid with a 16-carbon backbone and a PEG linker connected to a terminal NHS ester. The NHS ester can be used to label the primary amines (-NH2) of proteins, amine-modified oligonucleotides, and other amine-containing molecules. The hydrophilic PEG spacer increases solubility in aqueous media.
Catalog No. NC4036241 Supplier Broadpharm Supplier No. BP28231100MG
Palmitic acid-PEG4-NHS ester is a saturated long-chain fatty acid with a 16-carbon backbone and a PEG linker connected to a terminal NHS ester. The NHS ester can be used to label the primary amines (-NH2) of proteins, amine-modified oligonucleotides, and other amine-containing molecules. The hydrophilic PEG spacer increases solubility in aqueous media.