Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Palmitic acid Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111014684 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4684 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-4684 Supplier MilliporeSigma Supelco Supplier No. PHR11201G
Add to Cart
Add to Cart
Cleaning Products, Cosmetics, Food and Beverages, Personal Care and Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

CAS 57-10-3
Vapor Pressure 10 mmHg (210°C)
Density 0.852 g/mL (at 25°C)
Flash Point 113°C
Linear Formula CH3(CH2)14 COOH
MDL Number MFCD00002747
Melting Point 61°C to 62.5°C
Boiling Point 271.5°C (100 mmHg)
Synonym 1-Pentadecanecarboxylic acid; C16:0; Cetylic acid; Hexadecanoic acid; NSC 5030; PamOH
Type Certified Reference Material
Quantity 1 g
Formula Weight 256.42
Grade Certified Reference Material
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.