Learn More
Medchemexpress LLC Palmitic acid (sodium) | 408-35-5 | 278.41 | 10 G
SDP
Catalog No. 5000416254
Encompass_Preferred
Palmitic acid sodium is a long-chain saturated fatty acid commonly found in both animals and plants. It can induce the expression of glucose-regulated protein 78 (GRP78) and CCAAT/enhancer binding protein homologous protein (CHOP) in mouse granulosa cells. It is also used to establish a cell steatosis model.
  • Commonly found in animals and plants
  • Induces expression of glucose-regulated protein 78 (GRP78) and CCAAT/enhancer binding protein homologous protein (CHOP)
  • Used to establish a cell steatosis model
  • Increases mRNA levels of Notch1, 2 and 4 in hepatic cell lines
  • Store at 4°C, sealed, away from moisture
  • In solvent, store at -80°C for 6 months or -20°C for 1 month
Catalog No. 50-004-16254 Supplier Medchemexpress LLC Supplier No. HYN0830B10G
Palmitic acid sodium is a long-chain saturated fatty acid commonly found in both animals and plants. It can induce the expression of glucose-regulated protein 78 (GRP78) and CCAAT/enhancer binding protein homologous protein (CHOP) in mouse granulosa cells. It is also used to establish a cell steatosis model.
  • Commonly found in animals and plants
  • Induces expression of glucose-regulated protein 78 (GRP78) and CCAAT/enhancer binding protein homologous protein (CHOP)
  • Used to establish a cell steatosis model
  • Increases mRNA levels of Notch1, 2 and 4 in hepatic cell lines
  • Store at 4°C, sealed, away from moisture
  • In solvent, store at -80°C for 6 months or -20°C for 1 month