Learn More
Medchemexpress LLC Palmitic acid sodium | 408-35-5 | 278.41 | 5 G
SDP
Catalog No. 5000416439
Encompass_Preferred
Palmitic acid sodium is a long-chain saturated fatty acid, naturally occurring in both animals and plants. This compound is known for its ability to induce the expression of glucose-regulated protein 78 (GRP78) and CCAAT/enhancer binding protein homologous protein (CHOP) in mouse granulosa cells. It is commonly utilized in research to establish cell steatosis models.
  • Induces expression of GRP78 and CHOP in mouse granulosa cells.
  • Used to establish a cell steatosis model.
  • Increases mRNA levels of Notch1, 2, and 4 in LX2, Huh7, and MIHA hepatic cell lines.
Catalog No. 50-004-16439 Supplier Medchemexpress LLC Supplier No. HYN0830B5G
Add to Cart
Add to Cart
Palmitic acid sodium is a long-chain saturated fatty acid, naturally occurring in both animals and plants. This compound is known for its ability to induce the expression of glucose-regulated protein 78 (GRP78) and CCAAT/enhancer binding protein homologous protein (CHOP) in mouse granulosa cells. It is commonly utilized in research to establish cell steatosis models.
  • Induces expression of GRP78 and CHOP in mouse granulosa cells.
  • Used to establish a cell steatosis model.
  • Increases mRNA levels of Notch1, 2, and 4 in LX2, Huh7, and MIHA hepatic cell lines.